Dimethyl-trans-1,2-cyclopropanedicarboxylate 95%

Lieferant: Apollo Scientific

OR923179-1G OR923179-5G
APOSOR923179-1GEA 43.5 EUR
APOSOR923179-1G APOSOR923179-5G
Dimethyl-trans-1,2-cyclopropanedicarboxylate 95%
Dimethyl-trans-1,2-cyclopropanedicarboxylate
Formel: C₇H₁₀O₄
Lagertemperatur: Raumtemperatur
MDL: MFCD00062806
CAS-Nummer: 826-35-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR