cis-2,6-Dimethylpiperidin 95%

Lieferant: Apollo Scientific

OR923199-25G
APOSOR923199-25GEA 65.4 EUR
APOSOR923199-25G
cis-2,6-Dimethylpiperidin 95%
cis-2,6-Dimethylpiperidin
Formel: C₇H₁₅N
Lagertemperatur: Raumtemperatur
MDL: MFCD00066304
CAS-Nummer: 766-17-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR