1H-Pyrrolo[2,3-b]pyridin-3-ylmethanol 95%

Lieferant: Apollo Scientific

Synonyme: (1H-Pyrrolo[2,3-b]pyridin-3-yl)methanol

OR923206-1G OR923206-250MG
APOSOR923206-1GEA 109 EUR
APOSOR923206-1G APOSOR923206-250MG
1H-Pyrrolo[2,3-b]pyridin-3-ylmethanol 95%
1H-Pyrrolo[2,3-b]pyridin-3-ylmethanol
Formel: C₈H₈N₂O
Molecular Weight: 148.17 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11054203
CAS-Nummer: 1065100-83-5

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR