Ethylglycidylether 95%

Lieferant: Apollo Scientific

OR923221-100G OR923221-25G
APOSOR923221-100GEA 435 EUR
APOSOR923221-100G APOSOR923221-25G
Ethylglycidylether 95%
Ethylglycidylether
Formel: C₅H₁₀O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD00085264
CAS-Nummer: 4016-11-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR