2,3-Dicyanpyrazin 95%

Lieferant: Apollo Scientific

Synonyme: Pyrazine-2,3-dicarbonitrile

OR923278-25G
APOSOR923278-25GEA 198 EUR
APOSOR923278-25G
2,3-Dicyanpyrazin 95%
2,3-Dicyanopyrazine
Formel: C₆H₂N₄
Molecular Weight: 130,1069 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00059771
CAS-Nummer: 13481-25-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR