1-Isopropylpyrrolidin-3-ylamine dihydrochloride 95%

Lieferant: Apollo Scientific

Synonyme: 1-Isopropyl-pyrrolidin-3-ylamine dihydrochloride, 1-(Propan-2-yl)pyrrolidin-3-amine

OR923288-1G
APOSOR923288-1GEA 119 EUR
APOSOR923288-1G
1-Isopropylpyrrolidin-3-ylamine dihydrochloride 95%
1-Isopropylpyrrolidin-3-ylamine dihydrochloride
Formel: C₇H₁₈Cl₂N₂
Lagertemperatur: Raumtemperatur
MDL: MFCD08544449
CAS-Nummer: 19985-09-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR