1-Methyl-1H-pyrazolo[3,4-b]pyridine-3-carbonitrile 95%

Lieferant: Apollo Scientific

OR923320-1G
APOSOR923320-1GEA 487 EUR
APOSOR923320-1G
1-Methyl-1H-pyrazolo[3,4-b]pyridine-3-carbonitrile 95%
1-Methyl-1H-pyrazolo[3,4-b]pyridine-3-carbonitrile
Formel: C₈H₆N₄
Molecular Weight: 158,16 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18259208
CAS-Nummer: 1256795-04-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR