5-tert-Butyl-2-(chloromethyl)oxazole 95%
Lieferant: Apollo Scientific
OR923332-1G
APOSOR923332-1GEA
538
EUR
APOSOR923332-1G
5-tert-Butyl-2-(chloromethyl)oxazole 95%
5-tert-Butyl-2-(chloromethyl)oxazole
Formel:
C₈H₁₂ClNO Lagertemperatur: Raumtemperatur |
MDL:
MFCD08236738 CAS-Nummer: 224441-73-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...