2-Cyano-3-methyl-2-butenoic acid 95%

Lieferant: Apollo Scientific

OR923367-1G
APOSOR923367-1GEA 243 EUR
APOSOR923367-1G
2-Cyano-3-methyl-2-butenoic acid 95%
2-Cyano-3-methyl-2-butenoic acid
Formel: C₆H₇NO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD00192788
CAS-Nummer: 759-21-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR