4-Methoxy-3,3-dimethyl-4-oxobutanoic acid 95%

Lieferant: Apollo Scientific

OR923385-5G
APOSOR923385-5GEA 178 EUR
APOSOR923385-5G
4-Methoxy-3,3-dimethyl-4-oxobutanoic acid 95%
4-Methoxy-3,3-dimethyl-4-oxobutanoic acid
Formel: C₇H₁₂O₄
Molecular Weight: 160,17 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11617811
CAS-Nummer: 32980-26-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR