5,6-Dimethylpyridazin-3(2H)-one 95%

Lieferant: Apollo Scientific

OR923397-1G
APOSOR923397-1GEA 487 EUR
APOSOR923397-1G
5,6-Dimethylpyridazin-3(2H)-one 95%
5,6-Dimethylpyridazin-3(2H)-one
Formel: C₆H₈N₂O
Molecular Weight: 124,141 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD16871377
CAS-Nummer: 100114-19-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR