(S)-(-)-2-Methoxypropionic acid 97%

Lieferant: Apollo Scientific

OR923407-10G OR923407-1G OR923407-5G OR923407-25G
APOSOR923407-10GEA 0 EUR
APOSOR923407-10G APOSOR923407-1G APOSOR923407-5G APOSOR923407-25G
(S)-(-)-2-Methoxypropionic acid 97%
(S)-(-)-2-Methoxypropionic acid
Formel: C₄H₈O₃
Lagertemperatur: Raumtemperatur
MDL: MFCD01632589
CAS-Nummer: 23953-00-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR