2-(2-Methyl-1H-imidazol-1-yl)benzaldehyde 95%

Lieferant: Apollo Scientific

OR923428-1G OR923428-5G
APOSOR923428-1GEA 203 EUR
APOSOR923428-1G APOSOR923428-5G
2-(2-Methyl-1H-imidazol-1-yl)benzaldehyde 95%
2-(2-Methyl-1H-imidazol-1-yl)benzaldehyde
Formel: C₁₁H₁₀N₂O
Lagertemperatur: Raumtemperatur
MDL: MFCD05864668
CAS-Nummer: 914348-86-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR