Ethyl-o-toluat 95%
Lieferant: Apollo Scientific
Synonyme:
o-Toluylsäureethylester, 2-Tolylsäureethylester, Ethyl-2-methylbenzoat, 2-Methylbenzoesäureethylester, 2-Toluylsäureethylester
OR923435-100G
APOSOR923435-100GEA
95.7
EUR
APOSOR923435-100G
Ethyl-o-toluat 95%
Ethyl-o-toluat
Formel:
C₁₀H₁₂O₂ Molecular Weight: 164,2 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00009112 CAS-Nummer: 87-24-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...