3-[(Phenylmethyl)amino]-3-oxetanecarbonitrile 95%

Lieferant: Apollo Scientific

OR923438-1G
APOSOR923438-1GEA 609 EUR
APOSOR923438-1G
3-[(Phenylmethyl)amino]-3-oxetanecarbonitrile 95%
3-[(Phenylmethyl)amino]-3-oxetanecarbonitrile
Formel: C₁₁H₁₂N₂O
Lagertemperatur: Raumtemperatur
MDL: MFCD20687879
CAS-Nummer: 138650-20-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR