Ethyl tetrahydro-1H-pyrrolizine-7a(5H)-acetate 95%
Lieferant: Apollo Scientific
OR923487-1G
APOSOR923487-1GEA
364
EUR
APOSOR923487-1G
Ethyl tetrahydro-1H-pyrrolizine-7a(5H)-acetate 95%
Ethyl tetrahydro-1H-pyrrolizine-7a(5H)-acetate
Formel:
C₁₁H₁₉NO₂ Molecular Weight: 197,274 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD20487986 CAS-Nummer: 88069-56-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...