3-(1H-1,2,4-Triazol-1-yl)benzaldehyd 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-(1,2,4-Triazol-1-yl)benzaldehyde

OR9235-250MG
APOSOR9235-250MGEA 34.8 EUR
APOSOR9235-250MG
3-(1H-1,2,4-Triazol-1-yl)benzaldehyd 97%
3-(1H-1,2,4-Triazol-1-yl)benzaldehyd
Formel: C₉H₇N₃O
Molecular Weight: 173,1714 g/mol
Schmelzpunkt: 114,5…115.5 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD08271896
CAS-Nummer: 868755-54-8
UN: 1325
ADR: 4.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR