Cyclohexane-1,4-dicarbonitrile 95%

Lieferant: Apollo Scientific

OR923541-1G OR923541-5G
APOSOR923541-1GEA 230 EUR
APOSOR923541-1G APOSOR923541-5G
Cyclohexane-1,4-dicarbonitrile 95%
Cyclohexane-1,4-dicarbonitrile
Formel: C₈H₁₀N₂
Schmelzpunkt: 57…61 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00021283
CAS-Nummer: 10534-13-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR