2-Cyclohexylethanol 95%

Lieferant: Apollo Scientific

Synonyme: (2-Hydroxyethyl)cyclohexan

OR923566-100G
APOSOR923566-100GEA 109 EUR
APOSOR923566-100G
2-Cyclohexylethanol 95%
2-Cyclohexylethanol
Formel: C₈H₁₆O
Lagertemperatur: Raumtemperatur
MDL: MFCD00001525
CAS-Nummer: 4442-79-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR