4-(1-Ethylpropyl)pyridin 95%
Lieferant: Apollo Scientific
Synonyme:
4-(1-Ethylpropyl)pyridine, 4-(3-Pentyl)pyridine, 3-(4-Pyridyl)pentane
OR923607-1G
OR923607-25G
OR923607-5G
APOSOR923607-1GEA
139
EUR
APOSOR923607-1G
APOSOR923607-25G
APOSOR923607-5G
4-(1-Ethylpropyl)pyridin 95%
4-(1-Ethylpropyl)pyridine
Formel:
C₁₀H₁₅N Molecular Weight: 149.24 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00059788 CAS-Nummer: 35182-51-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...