5-Cyclopropyl-1,3,4-oxadiazol-2-amine 95%
Lieferant: Apollo Scientific
OR923615-1G
APOSOR923615-1GEA
125
EUR
APOSOR923615-1G
5-Cyclopropyl-1,3,4-oxadiazol-2-amine 95%
5-Cyclopropyl-1,3,4-oxadiazol-2-amine
Formel:
C₅H₇N₃O Lagertemperatur: Raumtemperatur |
MDL:
MFCD09041179 CAS-Nummer: 89179-60-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...