3-Bromo-4-methyl-1H-pyrazole

Lieferant: Apollo Scientific
Achtung

OR923657-1G OR923657-250MG OR923657-5G
APOSOR923657-1GEA 132 EUR
APOSOR923657-1G APOSOR923657-250MG APOSOR923657-5G
3-Bromo-4-methyl-1H-pyrazole
3-Bromo-4-methyl-1H-pyrazole
Formel: C₄H₅BrN₂
Lagertemperatur: Raumtemperatur
MDL: MFCD12755927
CAS-Nummer: 5932-20-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR