1-(2-Chlorophenyl)-2-methylpropan-2-ol 95%
Lieferant: Apollo Scientific
OR923685-1G
APOSOR923685-1GEA
184
EUR
APOSOR923685-1G
1-(2-Chlorophenyl)-2-methylpropan-2-ol 95%
1-(2-Chlorophenyl)-2-methylpropan-2-ol
Formel:
C₁₀H₁₃ClO Molecular Weight: 184,663 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD11610689 CAS-Nummer: 6256-31-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...