5-(Fur-2-yl)-1-methyl-1H-pyrazole-3-carboxaldehyde 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-Formyl-5-(fur-2-yl)-1-methyl-1H-pyrazole

OR9237-1G
APOSOR9237-1GEA 314 EUR
APOSOR9237-1G
5-(Fur-2-yl)-1-methyl-1H-pyrazole-3-carboxaldehyde 97%
5-(Fur-2-yl)-1-methyl-1H-pyrazole-3-carboxaldehyde
Formel: C₉H₈N₂O₂
Molecular Weight: 176,172 g/mol
Schmelzpunkt: 62…64 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD08271947
CAS-Nummer: 876728-40-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR