2-Cyclopentylcyclopentanon 95%

Lieferant: Apollo Scientific

Synonyme: [1,1'-Bi(cyclopentan)]-2-one

OR923715-25G
APOSOR923715-25GEA 87 EUR
APOSOR923715-25G
2-Cyclopentylcyclopentanon 95%
2-Cyclopentylcyclopentanone
Formel: C₁₀H₁₆O
Molecular Weight: 152.24 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00051419
CAS-Nummer: 4884-24-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR