Butyl-2-chloroethyl ether 95%

Lieferant: Apollo Scientific

OR923720-25G OR923720-5G
APOSOR923720-25GEA 0 EUR
APOSOR923720-25G APOSOR923720-5G
Butyl-2-chloroethyl ether 95%
Butyl-2-chloroethyl ether
Formel: C₆H₁₃ClO
Lagertemperatur: Raumtemperatur
MDL: MFCD00059051
CAS-Nummer: 10503-96-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR