-Cyclohexen-1-carbonitril 95%

Lieferant: Apollo Scientific

Synonyme: 1,2,3,6-Tetrahydrobenzonitrile, 3-Cyclohexene-1-carbonitrile

OR923732-5G
APOSOR923732-5GEA 145 EUR
APOSOR923732-5G
-Cyclohexen-1-carbonitril 95%
-Cyclohexen-1-carbonitril
Formel: C₇H₉N
Molecular Weight: 107.16 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00013778
CAS-Nummer: 100-45-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR