1-Benzyl-1,2-dihydropyridin-2-one 95%

Lieferant: Apollo Scientific

OR923763-1G OR923763-5G
APOSOR923763-1GEA 328 EUR
APOSOR923763-1G APOSOR923763-5G
1-Benzyl-1,2-dihydropyridin-2-one 95%
1-Benzyl-1,2-dihydropyridin-2-one
Formel: C₁₂H₁₁NO
Lagertemperatur: Raumtemperatur
MDL: MFCD00462400
CAS-Nummer: 1753-62-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR