5-Hexensäure 98%

Lieferant: Apollo Scientific

Synonyme: 5-Hexenoicacid

OR923776-25G
APOSOR923776-25GEA 184 EUR
APOSOR923776-25G
5-Hexensäure 98%
5-Hexenoic acid
Formel: C₆H₁₀O₂
Molecular Weight: 114.14 g/mol
Siedepunkt: 107
Schmelzpunkt: -37
Lagertemperatur: Raumtemperatur
MDL: MFCD00046558
CAS-Nummer: 1577-22-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR