2,5-Dichlorbenzylalkohol 95%
Lieferant: Apollo Scientific
Synonyme:
(2,5-Dichlorophenyl)methanol
OR923844-25G
APOSOR923844-25GEA
92.8
EUR
APOSOR923844-25G
2,5-Dichlorbenzylalkohol 95%
2,5-Dichlorbenzylalkohol
Formel:
C₇H₆Cl₂O Molecular Weight: 177.03 g/mol Siedepunkt: 264 Schmelzpunkt: 80 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00004607 CAS-Nummer: 34145-05-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...