Methyl[2-(methylsulphanyl)ethyl]amine hydrochloride 95%

Lieferant: Apollo Scientific

OR923872-1G
APOSOR923872-1GEA 487 EUR
APOSOR923872-1G
Methyl[2-(methylsulphanyl)ethyl]amine hydrochloride 95%
Methyl[2-(methylsulphanyl)ethyl]amine hydrochloride
Formel: C₄H₁₁NS·ClH
Molecular Weight: 141,663 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD15202271
CAS-Nummer: 98021-13-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR