3,6-Dichloro-4-ethylpyridazine 95%
Lieferant: Apollo Scientific
OR923895-1G
APOSOR923895-1GEA
538
EUR
APOSOR923895-1G
3,6-Dichloro-4-ethylpyridazine 95%
3,6-Dichloro-4-ethylpyridazine
Formel:
C₆H₆Cl₂N₂ Molecular Weight: 177,031 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD17014877 CAS-Nummer: 107228-54-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...