1-Benzylpyridin-4(1H)-one 95%
Lieferant: Apollo Scientific
OR923908-1G
APOSOR923908-1GEA
574
EUR
APOSOR923908-1G
1-Benzylpyridin-4(1H)-one 95%
1-Benzylpyridin-4(1H)-one
Formel:
C₁₂H₁₁NO Molecular Weight: 185.222 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00564929 CAS-Nummer: 72359-37-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...