trans-4-Nitrozimtaldehyd 95%
Lieferant: Apollo Scientific
Synonyme:
trans-3-(4-Nitrophenyl)-2-propenal, (E)-3-(4-Nitrophenyl)acrylaldehyde, (E)-4-Nitrocinnamaldehyde, (E)-3-(4-Nitrophenyl)-2-propenal
OR923923-1G
APOSOR923923-1GEA
56.7
EUR
APOSOR923923-1G
trans-4-Nitrozimtaldehyd 95%
trans-4-Nitrozimtaldehyd
Formel:
C₉H₇NO₃ Molecular Weight: 177.16 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00007379 CAS-Nummer: 49678-08-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...