1-(2-Naphthyl)ethanol 95%

Lieferant: Apollo Scientific

Synonyme: alpha-Methyl-2-naphthalenemethanol, 2-(1-Hydroxyethyl)naphthalene

OR923927-25G
APOSOR923927-25GEA 81.2 EUR
APOSOR923927-25G
1-(2-Naphthyl)ethanol 95%
1-(2-Naphthyl)ethanol
Formel: C₁₂H₁₂O
Molecular Weight: 172.23 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00004111
CAS-Nummer: 7228-47-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR