Pyrido[3,2-d]pyrimidin-4(3H)-one 96%

Lieferant: Apollo Scientific

OR923956-1G
APOSOR923956-1GEA 94.4 EUR
APOSOR923956-1G
Pyrido[3,2-d]pyrimidin-4(3H)-one 96%
Pyrido[3,2-d]pyrimidin-4(3H)-one
Formel: C₇H₅N₃O
Molecular Weight: 147.134 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02645992
CAS-Nummer: 37538-67-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR