4-Bromo-6-chloropyridin-2-amine 95+%
Lieferant: Apollo Scientific
|
Achtung
|
OR923960-250MG
APOSOR923960-250MGEA
84.1
EUR
APOSOR923960-250MG
4-Bromo-6-chloropyridin-2-amine 95+%
4-Bromo-6-chloropyridin-2-amine
Formel:
C₅H₄BrClN₂ Molecular Weight: 207,456 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD14582020 CAS-Nummer: 1206250-19-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...