2,3-Dimethyl-1H-pyrrolo[2,3-b]pyridine 95%

Lieferant: Apollo Scientific

OR923961-1G
APOSOR923961-1GEA 299 EUR
APOSOR923961-1G
2,3-Dimethyl-1H-pyrrolo[2,3-b]pyridine 95%
2,3-Dimethyl-1H-pyrrolo[2,3-b]pyridine
Formel: C₉H₁₀N₂
Molecular Weight: 146.189 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00513469
CAS-Nummer: 10299-69-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR