7-Methyl-7h-pyrrolo[2,3-d]pyrimidin-4-amine 95%

Lieferant: Apollo Scientific

OR923975-1G
APOSOR923975-1GEA 647 EUR
APOSOR923975-1G
7-Methyl-7h-pyrrolo[2,3-d]pyrimidin-4-amine 95%
7-Methyl-7h-pyrrolo[2,3-d]pyrimidin-4-amine
Formel: C₇H₈N₄
Molecular Weight: 148,165 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD12924275
CAS-Nummer: 7752-54-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR