2,2-Dimethyl-1H-indene-1,3(2H)-dione 95%

Lieferant: Apollo Scientific

OR923993-1G OR923993-250MG
APOSOR923993-1GEA 310 EUR
APOSOR923993-1G APOSOR923993-250MG
2,2-Dimethyl-1H-indene-1,3(2H)-dione 95%
2,2-Dimethyl-1H-indene-1,3(2H)-dione
Formel: C₁₁H₁₀O₂
Molecular Weight: 174.196 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00144912
CAS-Nummer: 17190-77-1

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR