(S)-3-Amino-1-methyl-piperidine dihydrochloride 96%

Lieferant: Apollo Scientific

Synonyme: (S)-1-Methylpiperidin-3-amine dihydrochloride

OR924007-1G
APOSOR924007-1GEA 105 EUR
APOSOR924007-1G
(S)-3-Amino-1-methyl-piperidine dihydrochloride 96%
(S)-3-Amino-1-methyl-piperidine dihydrochloride
Formel: C₆H₁₄N₂·2ClH
Molecular Weight: 187,111 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD20926142
CAS-Nummer: 1157849-51-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR