2,3-Dichloro-6-hydroxybenzaldehyde 95%

Lieferant: Apollo Scientific

OR924028-1G
APOSOR924028-1GEA 632 EUR
APOSOR924028-1G
2,3-Dichloro-6-hydroxybenzaldehyde 95%
2,3-Dichloro-6-hydroxybenzaldehyde
Formel: C₇H₄Cl₂O₂
Molecular Weight: 191,011 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD23705585
CAS-Nummer: 51571-16-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR