Benzyl isoamyl ether 98%
Lieferant: Apollo Scientific
Synonyme:
Benzyl Isopentyl Ether
OR924030-25G
APOSOR924030-25GEA
183
EUR
APOSOR924030-25G
Benzyl isoamyl ether 98%
Benzyl isoamyl ether
Formel:
C₁₂H₁₈O Molecular Weight: 178,28 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00048371 CAS-Nummer: 122-73-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...