2-Amino-5-methoxybenzolsulfonsäure 97%
Lieferant: Apollo Scientific
|
Achtung
|
OR924096-500G
APOSOR924096-500GEA
426
EUR
APOSOR924096-500G
2-Amino-5-methoxybenzolsulfonsäure 97%
2-Amino-5-methoxybenzenesulphonic acid
Formel:
C₇H₉NO₄S Molecular Weight: 203.2157 g/mol |
MDL:
MFCD00035804 CAS-Nummer: 13244-33-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...