Aciclovir
Lieferant: Apollo Scientific
Synonyme:
1,9-dihydro-2-amino-9-((2-hydroxyethoxy)methyl)-6h-purin-6-on, Acycloguanosine, 2-amino-1,9-dihydro-9-((2-hydroxyethoxy)methyl)-6h-purin-6-one, Acyclovir, 9-[(2-Hydroxyethoxy)methyl]guanine, 2-Amino-1,9-dihydro-9-((2-hydroxyethoxy)methyl)-6H-purin-6-one, 2-Amino-1,9-dihydro-9-[(2-hydroxyethoxy)methyl]-6H-purin-6-one, Aciclovir, 2-Amino-9-((2-hydroxyethoxy)methyl)-1H-purin-6(9H)-one
OR9240T-100MG
APOSOR9240T-100MGEA
87.3
EUR
APOSOR9240T-100MG
Aciclovir
Acyclovir
Formel:
C₈H₁₁N₅O₃ Molecular Weight: 225,21 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00057880 CAS-Nummer: 59277-89-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...