4,4-Dimethyl-oxazolidin-2-one 95%

Lieferant: Apollo Scientific

Synonyme: 4,4-Dimethyloxazolidin-2-one

OR924113-1G
APOSOR924113-1GEA 84.1 EUR
APOSOR924113-1G
4,4-Dimethyl-oxazolidin-2-one 95%
4,4-Dimethyl-oxazolidin-2-one
Formel: C₅H₉NO₂
Molecular Weight: 115,13 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD04038702
CAS-Nummer: 26654-39-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR