8-Oxabicyclo[3.2.1]octan-3-one 95%

Lieferant: Apollo Scientific

OR924122-1G
APOSOR924122-1GEA 97.3 EUR
APOSOR924122-1G
8-Oxabicyclo[3.2.1]octan-3-one 95%
8-Oxabicyclo[3.2.1]octan-3-one
Formel: C₇H₁₀O₂
Molecular Weight: 126,153 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD23106461
CAS-Nummer: 77745-32-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR