1-Methyl-1H-pyrazole-5-carbonitrile 95%

Lieferant: Apollo Scientific

OR924140-1G
APOSOR924140-1GEA 69.6 EUR
APOSOR924140-1G
1-Methyl-1H-pyrazole-5-carbonitrile 95%
1-Methyl-1H-pyrazole-5-carbonitrile
Formel: C₅H₅N₃
Molecular Weight: 107.12 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09701708
CAS-Nummer: 66121-72-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR