(S)-2-N-Boc-propane-1,2-diamine 97%

Lieferant: Apollo Scientific

OR924219-1G
APOSOR924219-1GEA 112 EUR
APOSOR924219-1G
(S)-2-N-Boc-propane-1,2-diamine 97%
(S)-2-N-Boc-propane-1,2-diamine
Formel: C₈H₁₈N₂O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD08726031
CAS-Nummer: 146552-71-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR