1,4-Dimethyl-5-nitroimidazole 95%

Lieferant: Apollo Scientific

OR924230-1G
APOSOR924230-1GEA 248 EUR
APOSOR924230-1G
1,4-Dimethyl-5-nitroimidazole 95%
1,4-Dimethyl-5-nitroimidazole
Formel: C₅H₇N₃O₂
Molecular Weight: 141,128 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01721570
CAS-Nummer: 57658-79-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR